Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(4-(4-(4-Methoxyphenoxy)-2,6-dimethylphenyl)thiazol-2-yl)isonicotinamide ≥98%

Supplier: AMBEED, INC
Warning

A310152-5MG A310152-10MG
77748-606EA 70 USD InStock
77748-606 77748-605
N-(4-(4-(4-Methoxyphenoxy)-2,6-dimethylphenyl)thiazol-2-yl)isonicotinamide ≥98%
N-(4-(4-(4-Methoxyphenoxy)-2,6-dimethylphenyl)thiazol-2-yl)isonicotinamide
Precaución: for Research Use Only


Formula: C₂₄H₂₁N₃O₃S
MDL Number: MFCD28348371
CAS Number: 1334921-03-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty