Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methanesulfonylamino-acetic acid ≥98%

Supplier: AMBEED, INC
Warning

A242817-100MG A242817-5G
77672-170EA 18 USD InStock
77672-170 77672-172
Methanesulfonylamino-acetic acid ≥98%
Methanesulfonylamino-acetic acid
Precaución: for Research Use Only


Formula: C₃H₇NO₄S
MDL Number: MFCD00069044
CAS Number: 35688-18-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty