Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,4-Dihydroxy-6-pentylbenzoic acid ≥95%

Supplier: AMBEED, INC
Warning

A318844-250MG A318844-100MG A318844-1G
77593-890EA 80.98 USD InStock
77593-890 77685-912 77685-913
2,4-Dihydroxy-6-pentylbenzoic acid ≥95%
2,4-Dihydroxy-6-pentylbenzoic acid
Precaución: for Research Use Only


Formula: C₁₂H₁₆O₄
MW: 224.25 g/mol
MDL Number: MFCD28383644
CAS Number: 491-72-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty