Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 4-((tert-butyldimethylsilyl)oxy)but-2-ynoate ≥96%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A325789-1G A325789-250MG
77683-971EA 141 USD InStock
77683-971 77683-972
Methyl 4-((tert-butyldimethylsilyl)oxy)but-2-ynoate ≥96%, Ambeed.Inc
Methyl 4-((tert-butyldimethylsilyl)oxy)but-2-ynoate
Precaución: For research use only.


Formula: C₁₁H₂₀O₃Si
MDL Number: MFCD06658479
CAS Number: 117968-51-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty