Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

HSF1A ≥98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A327751-10MG A327751-2MG A327751-5MG
77683-977EA 62 USD InStock
77683-977 77683-978 77683-979
HSF1A ≥98%, Ambeed.Inc
HSF1A
Precaución: For research use only.


Formula: C₂₁H₁₉N₃O₂S₂
MDL Number: MFCD22580417
CAS Number: 1196723-93-9
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty