Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nɑ-Acetyl-L-lysine ≥98%

Supplier: AMBEED, INC
Warning

A328373-250MG A328373-1G A328373-25G
77571-938EA 5 USD InStock
77571-938 77626-616 77626-618
Nɑ-Acetyl-L-lysine ≥98%
Nalpha-Acetyl-L-lysine
Precaución: for Research Use Only


Formula: C₈H₁₆N₂O₃
MDL Number: MFCD00008233
CAS Number: 1946-82-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty