Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fenchyl alcohol ≥95%

Supplier: AMBEED, INC
Warning

A343960-25G A343960-500G
77668-574EA 15 USD InStock
77668-574 77668-576
Fenchyl alcohol ≥95%
Fenchyl alcohol
Precaución: for Research Use Only


Formula: C₁₀H₁₈O
MW: 154.25 g/mol
MDL Number: MFCD00003760
CAS Number: 1632-73-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty