Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nonadecanoic acid ≥98%

Supplier: AMBEED, INC
Warning

A345910-100G A345910-25G
77668-586EA 299 USD InStock
77668-586 77668-588
Nonadecanoic acid ≥98%
Nonadecanoic acid
Precaución: for Research Use Only


Formula: C₁₉H₃₈O₂
MW: 298.51 g/mol
Boiling Pt: 297
MDL Number: MFCD00002754
CAS Number: 646-30-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty