Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Ethyl(3-((4-((3-(2-((3-fluorophenyl)amino)-2-oxoethyl)-1H-pyrazol-5-yl)amino)quinazolin-7-yl)oxy)propyl)amino)ethyl dihydrogen phosphate ≥98%

Supplier: AMBEED, INC
Warning

A363646-10MG A363646-5MG
77668-684EA 137 USD InStock
77668-684 77668-686
2-(Ethyl(3-((4-((3-(2-((3-fluorophenyl)amino)-2-oxoethyl)-1H-pyrazol-5-yl)amino)quinazolin-7-yl)oxy)propyl)amino)ethyl dihydrogen phosphate ≥98%
2-(Ethyl(3-((4-((3-(2-((3-fluorophenyl)amino)-2-oxoethyl)-1H-pyrazol-5-yl)amino)quinazolin-7-yl)oxy)propyl)amino)ethyl dihydrogen phosphate
Precaución: for Research Use Only


Formula: C₂₆H₃₁FN₇O₆P
MDL Number: MFCD15146330
CAS Number: 722543-31-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty