Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Chloro(2-{bis[3,5-bis(trifluoromethyl)phenyl]phosphino}-3,6-dimethoxy-2',4',6'-triisopropyl-1,1'-biphenyl)gold(I) ≥98%

Supplier: AMBEED, INC
Warning

A373092-100MG A373092-250MG
77748-746EA 188 USD InStock
77748-746 77748-747
Chloro(2-{bis[3,5-bis(trifluoromethyl)phenyl]phosphino}-3,6-dimethoxy-2',4',6'-triisopropyl-1,1'-biphenyl)gold(I) ≥98%
Chloro(2-{bis[3,5-bis(trifluoromethyl)phenyl]phosphino}-3,6-dimethoxy-2',4',6'-triisopropyl-1,1'-biphenyl)gold(I)
Precaución: for Research Use Only


Formula: C₃₉H₃₇AuClF₁₂O₂P
MDL Number: MFCD22200550
CAS Number: 1334547-76-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty