Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium Diphenylphosphinobenzene-3-sulfonate ≥90%

Supplier: AMBEED, INC
Danger

A378228-5G A378228-1G
77626-740EA 729.24 USD InStock
77626-740 77626-738
Sodium Diphenylphosphinobenzene-3-sulfonate ≥90%
Sodium Diphenylphosphinobenzene-3-sulfonate
Precaución: for Research Use Only


Formula: C₁₈H₁₅O₃PS
MW: 364.33 g/mol
MDL Number: MFCD00144157
CAS Number: 63995-75-5
UN: 3261
ADR: 8 Ⅲ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty