Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-(1H-Benzo[d]imidazol-1-yl)-N-(3-fluorophenyl)acetamido)-N-cyclohexyl-2-(o-tolyl)acetamide ≥98%

Supplier: AMBEED, INC
Warning

A376787-5MG A376787-2MG A376787-10MG
77746-754EA 52 USD InStock
77746-754 77746-753 77746-752
2-(2-(1H-Benzo[d]imidazol-1-yl)-N-(3-fluorophenyl)acetamido)-N-cyclohexyl-2-(o-tolyl)acetamide ≥98%
2-(2-(1H-Benzo[d]imidazol-1-yl)-N-(3-fluorophenyl)acetamido)-N-cyclohexyl-2-(o-tolyl)acetamide


Formula: C₃₀H₃₁FN₄O₂
MDL Number: MFCD28385853
CAS Number: 1355326-21-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty