Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Apramycin sulfate ≥550 µg/mg (dried basis)

Supplier: AMBEED, INC
Danger

A382039-5G A382039-250MG A382039-1G
77748-773EA 56 USD InStock
77748-773 77748-772 77748-771
Apramycin sulfate ≥550 µg/mg (dried basis)
Apramycin sulfate
Precaución: for Research Use Only


Formula: C₂₁H₄₁N₅O₁₁·H₂SO₄
MDL Number: MFCD06200257
CAS Number: 65710-07-8
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty