Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Ethoxybenzenesulfonyl chloride ≥98%

Supplier: AMBEED, INC
Danger

A387521-5G A387521-1G
77748-782EA 227 USD InStock
77748-782 77748-781
4-Ethoxybenzenesulfonyl chloride ≥98%
4-Ethoxybenzenesulfonyl chloride
Precaución: for Research Use Only


Formula: C₈H₉ClO₃S
MW: 220.68 g/mol
MDL Number: MFCD03147322
CAS Number: 1132-17-8
UN: 3261
ADR: 8 Ⅲ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty