Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2-((2,4-Dimethylphenyl)sulfinyl)phenyl)piperazine ≥97%

Supplier: AMBEED, INC
Danger

A402641-250MG A402641-100MG
77746-805EA 299 USD InStock
77746-805 77746-804
1-(2-((2,4-Dimethylphenyl)sulfinyl)phenyl)piperazine ≥97%
1-(2-((2,4-Dimethylphenyl)sulfinyl)phenyl)piperazine
Precaución: for Research Use Only


Formula: C₁₈H₂₂N₂OS
MDL Number: MFCD30543692
CAS Number: 1429908-35-9
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty