Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Linoelaidic acid ≥98%

Supplier: AMBEED, INC
Warning

A404888-100MG A404888-1G A404888-250MG
77601-784EA 118.9 USD InStock
77601-784 77668-850 77668-852
Linoelaidic acid ≥98%
Linoelaidic acid
Precaución: for Research Use Only


Formula: C₁₈H₃₂O₂
MDL Number: MFCD00004435
CAS Number: 506-21-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty