Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,2-Dimethyl-3,4-dihydro-2H-benzo[g]chromene-5,10-dione ≥98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A418258-1G A418258-250MG
77684-171EA 1341 USD InStock
77684-171 77684-172
2,2-Dimethyl-3,4-dihydro-2H-benzo[g]chromene-5,10-dione ≥98%, Ambeed.Inc
2,2-Dimethyl-3,4-dihydro-2H-benzo[g]chromene-5,10-dione
Precaución: For research use only.


Formula: C₁₅H₁₄O₃
MDL Number: MFCD00022287
CAS Number: 4707-33-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty