Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

((((S)-1-(4-Bromophenyl)ethyl)amino)(2,3-dioxo-1,2,3,4-tetrahydroquinoxalin-5-yl)methyl)phosphonic acid ≥98%

Supplier: AMBEED, INC
Warning

A432274-10MG A432274-5MG
77746-877EA 218 USD InStock
77746-877 77746-878
((((S)-1-(4-Bromophenyl)ethyl)amino)(2,3-dioxo-1,2,3,4-tetrahydroquinoxalin-5-yl)methyl)phosphonic acid ≥98%
((((S)-1-(4-Bromophenyl)ethyl)amino)(2,3-dioxo-1,2,3,4-tetrahydroquinoxalin-5-yl)methyl)phosphonic acid
Precaución: for Research Use Only


Formula: C₁₇H₁₇BrN₃O₅P
MDL Number: MFCD16628139
CAS Number: 459836-30-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty