Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bis[1,2-bis(diphenylphosphino)ethane]palladium(0) ≥98%

Supplier: AMBEED, INC
Warning

A433347-100MG A433347-1G A433347-5G
77668-928EA 16 USD InStock
77668-928 77668-930 77668-932
Bis[1,2-bis(diphenylphosphino)ethane]palladium(0) ≥98%
Bis[1,2-bis(diphenylphosphino)ethane]palladium(0)
Precaución: for Research Use Only


Formula: C₅₂H₄₈P₄Pd
MW: 903.27 g/mol
MDL Number: MFCD00009880
CAS Number: 31277-98-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty