Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Stearoyl ethanolamide ≥95%

Supplier: AMBEED, INC
Danger

A443524-1G A443524-25G A443524-5G
77668-982EA 109 USD InStock
77668-982 77668-984 77668-986
Stearoyl ethanolamide ≥95%
Stearoyl ethanolamide
Precaución: for Research Use Only


Formula: C₂₀H₄₁NO₂
MDL Number: MFCD00045971
CAS Number: 111-57-9
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty