Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Succinimidyl 7-Methoxycoumarin-3-carboxylate ≥98%

Supplier: AMBEED, INC
Warning

A446518-100MG A446518-1G A446518-250MG
77668-998EA 67 USD InStock
77668-998 77669-000 77669-002
N-Succinimidyl 7-Methoxycoumarin-3-carboxylate ≥98%
N-Succinimidyl 7-Methoxycoumarin-3-carboxylate
Precaución: for Research Use Only


Formula: C₁₅H₁₁NO₇
MW: 317.25 g/mol
MDL Number: MFCD00467593
CAS Number: 150321-92-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty