Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Z)-3-(Dimethylamino)propane-1,2-diyl dioleate ≥98%

Supplier: AMBEED, INC
Warning

A439003-100MG A439003-1G
77626-878EA 135.96 USD InStock
77626-878 77626-880
(Z)-3-(Dimethylamino)propane-1,2-diyl dioleate ≥98%
(Z)-3-(Dimethylamino)propane-1,2-diyl dioleate
Precaución: for Research Use Only


Formula: C₄₁H₇₇NO₄
MDL Number: MFCD01321074
CAS Number: 127512-29-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty