Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-6-bromo-5-methylbenzoic acid ≥97%

Supplier: AMBEED, INC
Warning

A451261-100MG A451261-250MG
77746-925EA 32 USD InStock
77746-925 77746-926
2-Amino-6-bromo-5-methylbenzoic acid ≥97%
2-Amino-6-bromo-5-methylbenzoic acid
Precaución: for Research Use Only


Formula: C₈H₈BrNO₂
MDL Number: MFCD01872212
CAS Number: 147149-85-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty