Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Hydroxyhexyl 4-methylbenzenesulfonate ≥95%

Supplier: AMBEED, INC
Warning

A458641-100MG A458641-1G A458641-250MG A458641-5G
77686-227EA 24 USD InStock
77686-227 77686-228 77686-229 77686-230
6-Hydroxyhexyl 4-methylbenzenesulfonate ≥95%
6-Hydroxyhexyl 4-methylbenzenesulfonate
Precaución: for Research Use Only


Formula: C₁₃H₂₀O₄S
MDL Number: MFCD00092068
CAS Number: 75351-33-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty