Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Decenoic acid ≥98% (by GC, titration analysis)

Supplier: AMBEED, INC
Warning

A460097-100G A460097-1G A460097-25G
77622-314EA 763.23 USD InStock
77622-314 77622-316 77622-318
4-Decenoic acid ≥98% (by GC, titration analysis)
4-Decenoic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₈O₂
MDL Number: MFCD03092963
CAS Number: 26303-90-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty