Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(E)-7-((1R,2R,3R)-3-Hydroxy-2-((3S,5S,E)-3-hydroxy-5-methylnon-1-en-1-yl)-5-oxocyclopentyl)hept-2-enoic acid ≥98%

Supplier: AMBEED, INC
Danger

A465257-10MG A465257-5MG
77669-090EA 295 USD InStock
77669-090 77669-092
(E)-7-((1R,2R,3R)-3-Hydroxy-2-((3S,5S,E)-3-hydroxy-5-methylnon-1-en-1-yl)-5-oxocyclopentyl)hept-2-enoic acid ≥98%
(E)-7-((1R,2R,3R)-3-Hydroxy-2-((3S,5S,E)-3-hydroxy-5-methylnon-1-en-1-yl)-5-oxocyclopentyl)hept-2-enoic acid
Precaución: for Research Use Only


Formula: C₂₂H₃₆O₅
MW: 380.52 g/mol
MDL Number: MFCD00868275
CAS Number: 74397-12-9
UN: 2811
ADR: 6.1 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty