Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dibutyl phosphate ≥98%

Supplier: AMBEED, INC
Danger

A469988-25G A469988-100G
77594-482EA 19.48 USD InStock
77594-482 77626-938
Dibutyl phosphate ≥98%
Dibutyl phosphate
Precaución: for Research Use Only


Formula: C₈H₁₉O₄P
MW: 210.21 g/mol
MDL Number: MFCD00015245
CAS Number: 107-66-4
UN: 3265
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty