Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxytrityl alcohol ≥95%

Supplier: AMBEED, INC
Warning

A493324-1G A493324-250MG
77686-290EA 19 USD InStock
77686-290 77686-291
4-Methoxytrityl alcohol ≥95%
4-Methoxytrityl alcohol
Precaución: for Research Use Only


Formula: C₂₀H₁₈O₂
MDL Number: MFCD00087962
CAS Number: 847-83-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty