Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-D-Pen(pMeOBzl)-OH ≥98%

Supplier: AMBEED, INC
Warning

A496987-100MG A496987-250MG
77626-982EA 21.63 USD InStock
77626-982 77626-984
Boc-D-Pen(pMeOBzl)-OH ≥98%
Boc-D-Pen(pMeOBzl)-OH
Precaución: for Research Use Only


Formula: C₁₈H₂₇NO₅S
MW: 369.48 g/mol
MDL Number: MFCD00076973
CAS Number: 106306-57-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty