Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

10-Acetyl-3,7-dihydroxyphenoxazine ≥98%

Supplier: AMBEED, INC
Warning

A501086-50MG A501086-100MG
77602-160EA 124.03 USD InStock
77602-160 77747-024
10-Acetyl-3,7-dihydroxyphenoxazine ≥98%
10-Acetyl-3,7-dihydroxyphenoxazine
Precaución: for Research Use Only


Formula: C₁₄H₁₁NO₄
MDL Number: MFCD00467861
CAS Number: 119171-73-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty