Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cholic acid ≥96%

Supplier: AMBEED, INC
Warning

A502353-100MG A502353-500G
77626-996EA 5.15 USD InStock
77626-996 77626-998
Cholic acid ≥96%
Cholic acid
Precaución: for Research Use Only


Formula: C₂₄H₄₀O₅
MW: 408.58
Boiling Pt: 545
Melting Pt: 199
MDL Number: MFCD00003672
CAS Number: 81-25-4
Merck Index: 13,02223
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty