Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Palmitelaidic acid ≥98%

Supplier: AMBEED, INC
Warning

A505831-10MG A505831-5MG
77669-262EA 46 USD InStock
77669-262 77669-264
Palmitelaidic acid ≥98%
Palmitelaidic acid
Precaución: for Research Use Only


MW: 254.43 g/mol
MDL Number: MFCD00067230
CAS Number: 10030-73-6
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty