Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(tert-Butylimino)-3-isopropyl-5-phenyl-1,3,5-thiadiazinan-4-one ≥98%

Supplier: AMBEED, INC
Danger

A518628-100MG A518628-10MG A518628-50MG
77673-372EA 53 USD InStock
77673-372 77673-374 77673-376
2-(tert-Butylimino)-3-isopropyl-5-phenyl-1,3,5-thiadiazinan-4-one ≥98%
2-(tert-Butylimino)-3-isopropyl-5-phenyl-1,3,5-thiadiazinan-4-one
Precaución: for Research Use Only


Formula: C₁₆H₂₃N₃OS
MDL Number: MFCD01681899
CAS Number: 69327-76-0
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty