Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

7-(2,3-Dihydroxypropyl)theophylline ≥98%

Supplier: AMBEED, INC
Warning

A520792-1000G A520792-100MG
77627-052EA 237.93 USD InStock
77627-052 77627-054
7-(2,3-Dihydroxypropyl)theophylline ≥98%
7-(2,3-Dihydroxypropyl)theophylline
Precaución: for Research Use Only


Formula: C₁₀H₁₄N₄O₄
MDL Number: MFCD00005759
CAS Number: 479-18-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty