Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

JZL-184 ≥98%

Supplier: AMBEED, INC
Danger

A513618-100MG A513618-10MG A513618-250MG
77686-339EA 88 USD InStock
77686-339 77686-340 77686-341
JZL-184 ≥98%
JZL-184
Precaución: for Research Use Only


Formula: C₂₇H₂₄N₂O₉
MDL Number: MFCD11976911
CAS Number: 1101854-58-3
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty