Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Bicyclo[2.2.1]heptan-2-ol ≥95%

Supplier: AMBEED, INC
Warning

A514348-250MG A514348-5G
77627-022EA 15.45 USD InStock
77627-022 77627-024
Bicyclo[2.2.1]heptan-2-ol ≥95%
Bicyclo[2.2.1]heptan-2-ol
Precaución: for Research Use Only


Formula: C₇H₁₂O
MW: 112.17 g/mol
MDL Number: MFCD00167747
CAS Number: 1632-68-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty