Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium hippurate ≥98%

Supplier: AMBEED, INC
Warning

A525519-100G A525519-25G A525519-500G
77669-324EA 64 USD InStock
77669-324 77669-326 77669-328
Sodium hippurate ≥98%
Sodium hippurate
Precaución: for Research Use Only


Formula: C₉H₈NNaO₃
MW: 201.16 g/mol
MDL Number: MFCD00002693
CAS Number: 532-94-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty