Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 1H-pyrazolo[3,4-c]pyridine-3-carboxylate ≥98%

Supplier: AMBEED, INC
Warning

A534579-100MG A534579-1G
77669-368EA 83 USD InStock
77669-368 77669-370
Ethyl 1H-pyrazolo[3,4-c]pyridine-3-carboxylate ≥98%
Ethyl 1H-pyrazolo[3,4-c]pyridine-3-carboxylate
Precaución: for Research Use Only


Formula: C₉H₉N₃O₂
MDL Number: MFCD10566315
CAS Number: 1053656-33-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty