Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Spiro[2.3]hexan-5-one ≥98%

Supplier: AMBEED, INC
Danger

A545332-100MG A545332-250MG
77622-358EA 57.68 USD InStock
77622-358 77622-360
Spiro[2.3]hexan-5-one ≥98%
Spiro[2.3]hexan-5-one
Precaución: for Research Use Only


Formula: C₆H₈O
MDL Number: MFCD23884029
CAS Number: 20061-22-7
UN: 1224
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty