Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Penicillin V potassium salt ≥98%

Supplier: AMBEED, INC
Warning

A548724-25G A548724-100G A548724-250MG A548724-500G A548724-5G
77602-320EA 69.7 USD InStock
77602-320 77669-424 77669-426 77669-428 77669-430
Penicillin V potassium salt ≥98%
Penicillin V potassium salt
Precaución: for Research Use Only


Formula: C₁₆H₁₇KN₂O₅S
MDL Number: MFCD00051771
CAS Number: 132-98-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty