Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Allantoic acid ≥95%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A557594-10MG A557594-5MG
77684-406EA 49 USD InStock
77684-406 77684-407
Allantoic acid ≥95%, Ambeed.Inc
Allantoic acid


Formula: C₄H₈N₄O₄
MDL Number: MFCD00007949
CAS Number: 99-16-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty