Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sisomicin sulphate Salt ≥98%

Supplier: AMBEED, INC
Danger

A558121-1MG A558121-50MG
77669-476EA 10 USD InStock
77669-476 77669-478
Sisomicin sulphate Salt ≥98%
Sisomicin sulphate Salt
Precaución: for Research Use Only


Formula: (C₁₉H₃₇N₅O₇)₂·5H₂SO₄
MW: 1385.4 g/mol
MDL Number: MFCD00058308
CAS Number: 53179-09-2
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty