Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Choline bromide 98% (by titrimetric analysis)

Supplier: AMBEED, INC
Warning

A590953-100G A590953-5G
77617-486EA 150.38 USD InStock
77617-486 77749-216
Choline bromide 98% (by titrimetric analysis)
Choline bromide
Precaución: for Research Use Only


Formula: C₅H₁₄BrNO
MDL Number: MFCD00054244
CAS Number: 1927-06-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty