Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Snap ≥98%

Supplier: AMBEED, INC
Warning

A594207-1G A594207-250MG
77686-507EA 188 USD InStock
77686-507 77686-508
Snap ≥98%
Snap
Precaución: for Research Use Only


Formula: C₇H₁₂N₂O₄S
MDL Number: MFCD00272624
CAS Number: 67776-06-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty