Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(4-(2-(Dimethylamino)ethoxy)benzyl)-3,4,5-trimethoxybenzamide ≥95%

Supplier: AMBEED, INC
Warning

A585716-100MG A585716-1G A585716-250MG
77669-540EA 63 USD InStock
77669-540 77669-542 77669-544
N-(4-(2-(Dimethylamino)ethoxy)benzyl)-3,4,5-trimethoxybenzamide ≥95%
N-(4-(2-(Dimethylamino)ethoxy)benzyl)-3,4,5-trimethoxybenzamide
Precaución: for Research Use Only


Formula: C₂₁H₂₈N₂O₅
MDL Number: MFCD00599601
CAS Number: 138-56-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty