Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Aminophylline 98+%

Supplier: AMBEED, INC
Danger

A594330-100MG A594330-5G
77627-226EA 5.15 USD InStock
77627-226 77627-228
Aminophylline 98+%
Aminophylline
Precaución: for Research Use Only


Formula: C₇H₈N₄O₂
MW: 420.43 g/mol
MDL Number: MFCD00013221
CAS Number: 317-34-0
UN: 2811
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty