Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium 5-(cyclohex-1-en-1-yl)-3-(trans-N-(trans-4-methoxycyclohexyl)-4-methylcyclohexane-1-carboxamido)thiophene-2-carboxylate ≥98%

Supplier: AMBEED, INC
Warning

A595198-10MG A595198-5MG
77686-514EA 198 USD InStock
77686-514 77686-515
Potassium 5-(cyclohex-1-en-1-yl)-3-(trans-N-(trans-4-methoxycyclohexyl)-4-methylcyclohexane-1-carboxamido)thiophene-2-carboxylate ≥98%
Potassium 5-(cyclohex-1-en-1-yl)-3-(trans-N-(trans-4-methoxycyclohexyl)-4-methylcyclohexane-1-carboxamido)thiophene-2-carboxylate
Precaución: for Research Use Only


Formula: C₂₆H₃₆KNO₄S
MW: 497.73 g/mol
MDL Number: MFCD25541650
CAS Number: 1200133-34-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty