Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dodecyl-β-D-glucopyranoside ≥98%, Ambeed.Inc

Supplier: AMBEED, INC
Warning

A607945-100MG A607945-250MG A607945-5G
77684-478EA 11 USD InStock
77684-478 77684-479 77684-480
Dodecyl-β-D-glucopyranoside ≥98%, Ambeed.Inc
Dodecyl-β-D-glucopyranoside
Precaución: For research use only.


Formula: C₁₈H₃₆O₆
MDL Number: MFCD00063298
CAS Number: 59122-55-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty