Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium bis(oxalato)oxotitanate(IV) dihydrate ≥98%

Supplier: AMBEED, INC
Danger

A609911-100G A609911-1000G
77594-868EA 8.2 USD InStock
77594-868 77686-539
Potassium bis(oxalato)oxotitanate(IV) dihydrate ≥98%
Potassium bis(oxalato)oxotitanate(IV) dihydrate
Precaución: for Research Use Only


Formula: K₂[TiO(C₂O₄)₂]·2H₂O
MW: 354.10 g/mol (318.10 anhy)
MDL Number: MFCD00150592
CAS Number: 14402-67-6
UN: 3288
ADR: 6.1 Ⅲ
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty