Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(+)-1,2-Bis[(2S,5S)-2,5-diethylphospholano]benzene(cyclooctadiene)rhodium(I) trifluoromethanesulfonate ≥98%

Supplier: AMBEED, INC
Warning

A611505-100MG A611505-250MG A611505-50MG
77686-542EA 328 USD InStock
77686-542 77686-543 77686-544
(+)-1,2-Bis[(2S,5S)-2,5-diethylphospholano]benzene(cyclooctadiene)rhodium(I) trifluoromethanesulfonate ≥98%
(+)-1,2-Bis[(2S,5S)-2,5-diethylphospholano]benzene(cyclooctadiene)rhodium(I) trifluoromethanesulfonate
Precaución: for Research Use Only


Formula: C₃₁H₄₈F₃O₃P₂RhS
MDL Number: MFCD00269861
CAS Number: 142184-30-3
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty